Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031378.3 | NP_113555.1 | 3 | 584 | ||
GenBank mRNA | AB042406.1 | - | 2 | 481 | ||
AK137662.1 | - | 3 | 585 |