Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006190.4 | NP_006181.1 | 3, 4 | 373 | ||
NR_033915.1 | 3, 4 | 373 | ||||
XM_006712555.3 | XP_006712618.1 | 3, 4 | 320 | |||
XM_011511252.2 | XP_011509554.1 | 3, 4 | 375 | |||
XM_011511253.2 | XP_011509555.1 | 3, 4 | 247 | |||
GenBank mRNA | AK302614.1 | - | 3, 4 | 345 | ||
AK316433.1 | - | 3, 4 | 274 | |||
BC014834.1 | - | 3, 4 | 316 | 2899929 | ||
BM470184.1 | - | 359 | ||||
DA111542.1 | - | 3, 4 | 373 | |||
U27459.1 | - | 3, 4 | 307 | |||
U40268.2 | - | 3, 4 | 288 |