Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002238.2 | NP_001002238.1 | 23 | 3387 | ||
NM_001002240.2 | NP_001002240.1 | 23 | 3360 | |||
NM_001002241.2 | NP_001002241.1 | 22 | 3660 | |||
NM_031387.3 | NP_113564.2 | 23 | 3520 | |||
GenBank mRNA | AB067571.1 | - | 22 | 3341 | ||
AB183526.1 | - | 23 | 3387 | |||
AB183527.1 | - | 23 | 3360 | |||
AB183528.1 | - | 22 | 3660 | |||
AB183529.1 | - | 22 | 3224 | |||
AF285591.1 | - | 19 | 2714 | |||
AK029806.1 | - | 23 | 3521 | |||
BC129954.1 | - | 18 | 2646 | 40081321 | ||
BC129955.1 | - | 18 | 2646 | 40081323 |