Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006191.2 | NP_006182.2 | 13 | 1956 | ||
GenBank mRNA | BC069786.1 | - | 13 | 1609 | 7262559 | |
U59435.1 | - | 13 | 1635 |