Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031406.3 | NP_113583.2 | 14 | 3066 | ||
GenBank mRNA | AF314957.1 | - | 13 | 2932 | ||
AK154118.1 | - | 14 | 3027 | |||
AK200107.1 | - | 1 | 17 | |||
BC022591.1 | - | 1 | 782 | |||
BC046982.1 | - | 14 | 2986 | 6478046 |