Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031863.3 | NP_114069.1 | 2 | 280 | ||
GenBank mRNA | AK011998.1 | - | 2 | 266 | ||
AK012179.1 | - | 2 | 281 | |||
AK020040.1 | - | 2 | 282 | |||
AK020761.1 | - | 2 | 776 | |||
AK076094.1 | - | 2 | 258 | |||
AK088680.1 | - | 111 | ||||
AK157569.1 | - | 2 | 308 | |||
BC018524.1 | - | 2 | 213 | 3588374 |