Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031871.1 | NP_114077.1 | 6 | 1210 | ||
GenBank mRNA | AF316996.1 | - | 6 | 1210 | ||
AK135322.1 | - | 6 | 1212 | |||
AK143922.1 | - | 6 | 1179 | |||
AK161571.1 | - | 3 | 358 | |||
BC005639.1 | - | 6 | 1191 | 3256858 |