Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031880.3 | NP_114086.3 | 8 | 1173 | ||
GenBank mRNA | AF307746.1 | - | 8 | 1173 | ||
AF332512.1 | - | 6 | 993 | |||
AK033440.1 | - | 3 | 544 | |||
AK076220.1 | - | 8 | 1179 | |||
AK167292.1 | - | 8 | 1174 | |||
BC029623.1 | - | 7 | 1160 | 4039780 | ||
BC055303.1 | - | 7 | 1077 | 6439288 |