Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114386.1 | NP_001107858.1 | 9 | 939 | ||
NM_031881.2 | NP_114087.2 | 8 | 988 | |||
GenBank mRNA | AF277232.1 | - | 8 | 621 | ||
AK004969.1 | - | 8 | 681 | |||
AK031842.1 | - | 8 | 669 | |||
AK042621.1 | - | 9 | 622 | |||
AK122283.1 | - | 8 | 791 | |||
AK163497.1 | - | 7 | 469 | |||
AK188093.1 | - | 4 | 322 | |||
AK200066.1 | - | 1 | 61 | |||
AK205175.1 | - | 4 | 321 | |||
BC039746.1 | - | 8 | 632 | 3157137 | ||
BC071210.1 | - | 8 | 609 | 4972426 | ||
BC082281.1 | - | 8 | 658 |