Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032000.2 | NP_114389.2 | 2 | 1108 | ||
GenBank mRNA | AF346836.1 | - | 3 | 1033 | ||
AK142355.1 | - | 2 | 778 | |||
BC037058.1 | - | 2 | 786 | 4951294 | ||
BC049857.1 | - | 4 | 674 | 4460789 | ||
BC099962.1 | - | 2 | 1073 | 30091008 |