Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032006.3 | NP_114395.1 | 7 | 1058 | ||
GenBank mRNA | AJ278462.1 | - | 7 | 1012 | ||
AK049552.1 | - | 7 | 1059 | |||
BC117756.2 | - | 7 | 1058 | 40090601 |