Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032396.3 | NP_115772.2 | 4 | 506 | ||
GenBank mRNA | AB059617.1 | - | 4 | 469 | ||
AK083565.1 | - | 2 | 114 | |||
AK141321.1 | - | 4 | 518 | |||
BC049771.1 | - | 4 | 430 | |||
BC082546.1 | - | 4 | 506 | 6855459 | ||
BC138464.1 | - | 4 | 514 | 8861484 | ||
BC138465.1 | - | 4 | 514 | 8861485 |