Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucnr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032400.2 | NP_115776.2 | 2 | 343 | ||
GenBank mRNA | AF295367.1 | - | 2 | 359 | ||
BB864882.1 | - | 2 | 347 | |||
BC096665.1 | - | 2 | 316 | 1365839 |
Set Membership: |
G-protein coupled receptor |