Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032465.2 | NP_115854.2 | 2 | 260 | ||
GenBank mRNA | AK040329.1 | - | 2 | 147 | ||
AK154076.1 | - | 2 | 230 | |||
AK156470.1 | - | 2 | 261 | |||
AY029156.1 | - | 2 | 121 | |||
BC052865.1 | - | 2 | 157 | 30072864 |