Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033321.3 | NP_201578.2 | 7, 8 | 828 | ||
GenBank mRNA | AF333331.1 | - | 7, 8 | 834 | ||
AK134662.1 | - | 7, 8 | 788 | |||
BC138189.1 | - | 7, 8 | 828 | 8861208 | ||
BC145181.1 | - | 7, 8 | 828 | 9053732 | ||
JX570808.1 | - | 6, 7 | 1091 |