Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033526.2 | NP_277068.1 | 11 | 2302 | ||
GenBank mRNA | AB040050.1 | - | 11 | 2302 | ||
AK077511.1 | - | 1 | 571 | |||
AK150211.1 | - | 11 | 2239 | |||
AK156515.1 | - | 11 | 2256 | |||
AK189891.1 | - | 1 | 86 | |||
AK190901.1 | - | 1 | 86 | |||
AK198811.1 | - | 1 | 86 | |||
AK202568.1 | - | 1 | 86 | |||
AK204386.1 | - | 1 | 86 | |||
BC017686.1 | - | 11 | 2264 | 4236887 |