Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242378.1 | NP_001229307.1 | 26 | 4865 | ||
NM_001242379.1 | NP_001229308.1 | 25 | 4775 | |||
NM_001242380.1 | NP_001229309.1 | 27 | 4904 | |||
NM_001242381.1 | NP_001229310.1 | 26 | 4811 | |||
NM_001242382.1 | NP_001229311.1 | 26 | 4862 | |||
NM_001242383.1 | NP_001229312.1 | 25 | 4772 | |||
NM_033552.3 | NP_291030.2 | 26 | 4814 | |||
GenBank mRNA | AB033759.1 | - | 26 | 4735 | ||
AK047801.1 | - | 1 | 823 | |||
AK220501.1 | - | 27 | 4869 | |||
BC039226.1 | - | 26 | 4763 | 4480841 |