Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033570.2 | NP_291048.2 | 1 | 1133 | ||
GenBank mRNA | AF216963.1 | - | 1 | 745 | ||
AK173209.1 | - | 1 | 1110 | |||
BC139799.1 | - | 1 | 734 | |||
GU390541.1 | - | 1 | 668 |