Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277891.1 | NP_001264820.1 | 4 | 456 | ||
NM_001277892.1 | NP_001264821.1 | 4 | 403 | |||
NM_001277893.1 | NP_001264822.1 | 4 | 456 | |||
NM_033571.3 | NP_291049.1 | 4 | 456 | |||
GenBank mRNA | AF367709.1 | - | 4 | 437 | ||
AF367710.1 | - | 4 | 403 | |||
AF367711.1 | - | 4 | 403 | |||
AK090036.1 | - | 4 | 447 | |||
AY029192.1 | - | 4 | 403 | |||
BC139092.1 | - | 4 | 456 | 8862114 | ||
BC139097.1 | - | 4 | 456 | 8862119 |