Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033572.2 | NP_291050.1 | 9 | 1173 | ||
GenBank mRNA | AF410456.1 | - | 9 | 1152 | ||
AK017733.1 | - | 9 | 1172 | |||
BC024714.1 | - | 9 | 1145 | 5134834 | ||
BC055335.1 | - | 9 | 1171 | 6412407 |