Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033582.2 | NP_291060.1 | 1 | 1456 | ||
GenBank mRNA | AK139349.1 | - | 1 | 194 | ||
AK139465.1 | - | 1 | 1080 | |||
AY013812.1 | - | 1 | 1002 | |||
BC064808.1 | - | 1 | 1456 | 6825007 |