Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033602.2 | NP_291080.2 | 4 | 740 | ||
GenBank mRNA | AF302504.1 | - | 4 | 669 | ||
AK030564.1 | - | 4 | 787 | |||
AK033815.1 | - | 1 | 46 | |||
AK045515.1 | - | 3 | 613 | |||
BC027062.1 | - | 3 | 579 | 5355240 |