Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242921.1 | NP_001229850.1 | 2 | 190 | ||
NM_006227.3 | NP_006218.1 | 4 | 563 | |||
NM_182676.2 | NP_872617.1 | 4 | 563 | |||
GenBank mRNA | AB076694.1 | - | 4 | 347 | ||
AF052157.1 | - | 1 | 65 | |||
AK092397.1 | - | 4 | 304 | |||
AK092523.1 | - | 4 | 351 | |||
AK097009.1 | - | 3 | 861 | |||
AK222690.1 | - | 4 | 351 | |||
AK289371.1 | - | 4 | 351 | |||
AK293150.1 | - | 2 | 106 | |||
BC005045.1 | - | 4 | 344 | 3510757 | ||
BC019847.1 | - | 4 | 345 | 4997011 | ||
BC019898.1 | - | 4 | 338 | 4992839 | ||
L26232.1 | - | 4 | 353 |