Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 19 | 3256 | ||
NM_033608.3 | NP_291086.2 | 19 | 3322 | |||
GenBank mRNA | AF317839.1 | - | 19 | 3322 | ||
AK078536.1 | - | 19 | 3251 | |||
AK145121.1 | - | 19 | 3235 | |||
BC023011.1 | - | 1 | 116 | |||
BC024487.1 | - | 1 | 94 | |||
BC034594.1 | - | 18 | 3085 | |||
BC040281.1 | - | 19 | 3282 | 5364073 |