Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172098.1 | NP_001165569.1 | 7 | 1117 | ||
NM_052973.2 | NP_443205.1 | 7 | 1117 | |||
GenBank mRNA | AF307777.1 | - | 7 | 1101 | ||
AK139183.1 | - | 7 | 1101 | |||
AK140447.1 | - | 7 | 1105 | |||
AK165908.1 | - | 7 | 801 | |||
BC138055.1 | - | 7 | 1068 | 8861074 | ||
BC145127.1 | - | 7 | 1068 | 9053675 | ||
EF685153.1 | - | 1 | 63 | |||
EF685154.1 | - | 2 | 190 | |||
EF685155.1 | - | 2 | 157 |