Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053093.1 | NP_444323.1 | 3 | 433 | ||
GenBank mRNA | AF235035.1 | - | 3 | 433 | ||
AF515827.1 | - | 3 | 267 | |||
BC119426.1 | - | 3 | 413 | 40039772 |