Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053100.2 | NP_444330.2 | 6 | 2438 | ||
GenBank mRNA | AF281047.1 | - | 6 | 1787 | ||
AK052763.1 | - | 6 | 1173 | |||
AK079847.1 | - | 4 | 877 | |||
AK155303.1 | - | 6 | 2385 | |||
BC060094.1 | - | 6 | 1668 | |||
BC082298.1 | - | 6 | 2313 | |||
BC152343.1 | - | 6 | 1774 | 40093266 | ||
BC152344.1 | - | 6 | 1774 | 40093269 |