Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053132.3 | NP_444362.3 | 1 | 2675 | ||
GenBank mRNA | AF326300.1 | - | 1 | 2471 | ||
AK029340.1 | - | 1 | 2654 | |||
AK034205.1 | - | 1 | 2677 | |||
AK043979.1 | - | 1 | 2634 | |||
AK046463.1 | - | 1 | 2658 |