Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006262.3 | NP_006253.2 | 9 | 1741 | ||
XM_005269025.1 | XP_005269082.1 | 9 | 1857 | |||
GenBank mRNA | AA460760.1 | - | 1 | 256 | ||
AK125587.1 | - | 9 | 3165 | |||
BC032703.1 | - | 9 | 1701 | 5247559 |