Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053139.3 | NP_444369.3 | 1 | 283 | ||
GenBank mRNA | AK008662.1 | - | 1 | 281 | ||
AK032156.1 | - | 1 | 286 | |||
AK083065.1 | - | 1 | 231 | |||
AY013775.1 | - | 1 | 97 | |||
BB622429.1 | - | 1 | 286 | |||
BC059821.1 | - | 1 | 260 | 6838626 |