Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 20 | 2942 | ||
XM_005264002.2 | XP_005264059.1 | 20 | 2977 | |||
XM_005264003.2 | XP_005264060.1 | 20 | 2905 | |||
XM_005264004.2 | XP_005264061.1 | 20 | 2905 | |||
XM_005264005.4 | XP_005264062.1 | 20 | 2965 | |||
XM_011511575.2 | XP_011509877.1 | 20 | 2880 | |||
XM_011511576.2 | XP_011509878.1 | 20 | 2905 | |||
XM_017004623.1 | XP_016860112.1 | 20 | 2905 | |||
XM_017004624.1 | XP_016860113.1 | 19 | 2810 | |||
GenBank mRNA | AB209483.1 | - | 17 | 2558 | ||
AK026993.1 | - | 4 | 371 | |||
D42063.1 | - | 20 | 2943 |