Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053146.2 | NP_444376.1 | 1 | 901 | ||
GenBank mRNA | AF326314.1 | - | 1 | 733 | ||
AK134846.1 | - | 901 | ||||
AY013783.1 | - | 1 | 733 | |||
BC138121.1 | - | 1 | 820 | 8861140 | ||
BC138122.1 | - | 1 | 820 | 8861141 |