Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053149.2 | NP_444379.1 | 4 | 1990 | ||
GenBank mRNA | AF269248.1 | - | 4 | 1990 | ||
AK077692.1 | - | 4 | 1833 | |||
AK156536.1 | - | 4 | 1918 | |||
AK171906.1 | - | 4 | 2045 | |||
AY699986.1 | - | 4 | 1822 |