Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053157.2 | NP_444387.1 | 3 | 367 | ||
GenBank mRNA | AF353993.1 | - | 3 | 341 | ||
AK009511.1 | - | 3 | 331 | |||
AK216092.1 | - | 3 | 245 | |||
BC089492.1 | - | 3 | 367 | 6703812 |