Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053164.3 | NP_444394.1 | 2 | 234 | ||
GenBank mRNA | AB049657.1 | - | 2 | 195 | ||
AK015357.1 | - | 2 | 215 | |||
AK076204.1 | - | 2 | 193 | |||
BC027676.1 | - | 2 | 174 | |||
BC048733.1 | - | 2 | 225 | 6512868 | ||
BC065812.1 | - | 2 | 181 |