Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135242.1 | NP_001128714.1 | 16 | 3424 | ||
NM_001258432.1 | NP_001245361.1 | 14 | 2910 | |||
NM_001258433.1 | NP_001245362.1 | 15 | 2921 | |||
NM_006096.3 | NP_006087.2 | 16 | 3027 | |||
XM_011516791.1 | XP_011515093.1 | 16 | 3361 | |||
XM_011516792.1 | XP_011515094.1 | 9 | 3260 | |||
GenBank mRNA | AF004162.1 | - | 15 | 2866 | ||
AF039944.1 | - | 1 | 243 | |||
AK091147.1 | - | 15 | 2870 | |||
AK091269.1 | - | 2 | 2761 | |||
AK095649.1 | - | 14 | 2763 | |||
AK096562.1 | - | 16 | 2862 | |||
AK124709.1 | - | 8 | 3934 | |||
AK126924.1 | - | 16 | 3706 | |||
AK129867.1 | - | 7 | 2301 | |||
AK130985.1 | - | 1 | 550 | |||
BC003175.1 | - | 16 | 2954 | 2957847 | ||
D87953.1 | - | 16 | 2960 | |||
X92845.1 | - | 16 | 2951 |