Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199272.1 | NP_001186201.1 | 1 | 414 | ||
NM_053187.3 | NP_444417.2 | 1 | 414 | |||
GenBank mRNA | AB052838.1 | - | 1 | 194 | ||
AF287893.1 | - | 1 | 330 | |||
AK030637.1 | - | 1 | 223 | |||
AK032613.1 | - | 1 | 220 | |||
AK171881.1 | - | 1 | 410 | |||
BC051171.1 | - | 1 | 365 | 6307523 |