Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1pr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053190.2 | NP_444420.1 | 3 | 1214 | ||
GenBank mRNA | AF327535.1 | - | 1 | 893 | ||
AK081126.1 | - | 1 | 893 | |||
BC012232.1 | - | 2 | 1139 | 4194177 |
Set Membership: |
G-protein coupled receptor |