Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053195.2 | NP_444425.1 | 17 | 2499 | ||
GenBank mRNA | BC005742.1 | - | 7 | 1629 | ||
BC017615.1 | - | 17 | 2499 | 4914291 |