Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178012.1 | NP_001171483.1 | 3 | 534 | ||
NM_001178013.1 | NP_001171484.1 | 3 | 531 | |||
NM_053197.4 | NP_444427.1 | 3 | 534 | |||
GenBank mRNA | AF325262.1 | - | 3 | 298 | ||
AK034514.1 | - | 3 | 251 | |||
AK089985.1 | - | 3 | 295 | |||
AK154274.1 | - | 3 | 304 | |||
BC012208.1 | - | 3 | 436 | 3985155 | ||
BY723942.1 | - | 3 | 535 |