Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098531.2 | NP_001092001.1 | 28 | 3532 | ||
NM_001098532.2 | NP_001092002.1 | 27 | 3296 | |||
NM_006105.5 | NP_006096.2 | 28 | 3066 | |||
XM_005268571.3 | XP_005268628.1 | 27 | 3042 | |||
XM_011537752.1 | XP_011536054.1 | 28 | 3549 | |||
XM_011537755.1 | XP_011536057.1 | 28 | 3111 | |||
XM_011537758.2 | XP_011536060.1 | 28 | 3161 | |||
GenBank mRNA | AK055502.1 | - | 10 | 1847 | ||
AK074444.1 | - | 12 | 2224 | |||
AK092448.1 | - | 28 | 3504 | |||
AK125545.1 | - | 24 | 2549 | |||
AK290230.1 | - | 29 | 3197 | |||
BC017728.1 | - | 28 | 3146 | 4414360 | ||
BC092404.1 | - | 28 | 3135 | 6181000 |