Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133350.1 | NP_579928.1 | 6 | 825 | ||
GenBank mRNA | AF468005.1 | - | 6 | 825 | ||
AK214734.1 | - | 1 | 57 | |||
BC048833.1 | - | 5 | 1387 | |||
BC057918.1 | - | 6 | 825 | 5134461 | ||
U51204.1 | - | 4 | 507 |