Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133721.2 | NP_598482.2 | 6 | 867 | ||
GenBank mRNA | AJ344342.1 | - | 6 | 870 | ||
AK153585.1 | - | 4 | 329 | |||
BC150824.1 | - | 6 | 773 | 9087735 |