Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113514.1 | NP_001106986.1 | 11 | 1160 | ||
NM_133721.2 | NP_598482.2 | 25 | 2981 | |||
GenBank mRNA | AJ344342.1 | - | 25 | 2984 | ||
AK083987.1 | - | 10 | 1000 | |||
AK139690.1 | - | 11 | 1148 | |||
AK153585.1 | - | 23 | 2443 |