Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 3 | 573 | ||
NM_001252460.1 | NP_001239389.1 | 3 | 628 | |||
NM_133769.3 | NP_598530.2 | 3 | 336 | |||
GenBank mRNA | AB093288.2 | - | 2 | 208 | ||
AF162472.1 | - | 2 | 196 | |||
AF334144.1 | - | 3 | 271 | |||
AK018280.1 | - | 3 | 266 | |||
AK030397.1 | - | 3 | 273 | |||
AK147224.1 | - | 3 | 574 | |||
AK147586.1 | - | 3 | 329 | |||
AK147632.1 | - | 3 | 270 | |||
AK147971.1 | - | 3 | 273 | |||
BC056974.1 | - | 3 | 284 | 6831770 | ||
CJ115523.1 | - | 3 | 337 |