Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252459.1 | NP_001239388.1 | 18, 19 | 2474 | ||
NM_001252460.1 | NP_001239389.1 | 18, 19 | 2529 | |||
NM_133769.3 | NP_598530.2 | 18, 19 | 2237 | |||
GenBank mRNA | AB093288.2 | - | 17, 18 | 2109 | ||
AF334144.1 | - | 18, 19 | 2172 | |||
AK030397.1 | - | 18, 19 | 2174 | |||
AK147224.1 | - | 18, 19 | 2475 | |||
AK147586.1 | - | 18, 19 | 2230 | |||
AK147632.1 | - | 18, 19 | 2171 | |||
AK147971.1 | - | 18, 19 | 2174 | |||
BC038080.1 | - | 1, 2 | 49 | |||
BC056974.1 | - | 18, 19 | 2185 | 6831770 |