Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133771.2 | NP_598532.1 | 9 | 1117 | ||
GenBank mRNA | AF363447.1 | - | 10 | 1179 | ||
AK002639.1 | - | 4 | 522 | |||
AK159560.1 | - | 9 | 1096 | |||
BC013819.1 | - | 9 | 1134 | 3489363 |