Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039878.2 | NP_001034967.1 | 12 | 1974 | ||
NM_133789.3 | NP_598550.2 | 12 | 1995 | |||
GenBank mRNA | AF414080.1 | - | 12 | 1995 | ||
AK136155.1 | - | 12 | 1291 | |||
AK154615.1 | - | 12 | 1524 | |||
AK161673.1 | - | 12 | 1545 | |||
BC004025.1 | - | 10 | 1108 | |||
BC080283.1 | - | 12 | 1561 | 6401782 |