Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833439L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252645.1 | NP_001239574.1 | 8 | 1490 | ||
NM_001252646.1 | NP_001239575.1 | 7 | 1383 | |||
NM_001252647.1 | NP_001239576.1 | 6 | 1204 | |||
NM_001252648.1 | NP_001239577.1 | 5 | 1077 | |||
NM_133797.4 | NP_598558.1 | 9 | 1578 | |||
GenBank mRNA | AK004845.1 | - | 9 | 1536 | ||
AK028697.1 | - | 9 | 1511 | |||
AK145025.1 | - | 9 | 1542 | |||
AK159945.1 | - | 9 | 1542 | |||
AK167616.1 | - | 9 | 1522 | |||
AK169399.1 | - | 9 | 1544 | |||
AK173126.1 | - | 9 | 1491 | |||
BC033445.1 | - | 9 | 1557 | 4222839 | ||
BC039775.1 | - | 9 | 1516 | 4239147 | ||
BC043040.1 | - | 9 | 1534 | 5701249 | ||
BC054417.1 | - | 9 | 1523 | 3969623 |
Gene Symbol: Ptrhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204912.1 | NP_001191841.1 | 2 | 2763 | ||
GenBank mRNA | BC067010.1 | - | 3 | 2680 |