Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006116.2 | NP_006107.1 | 7 | 798 | ||
NM_153497.2 | NP_705717.1 | 7 | 798 | |||
GenBank mRNA | AF425640.1 | - | 7 | 767 | ||
AK291668.1 | - | 7 | 756 | |||
AK296314.1 | - | 7 | 900 | |||
AK312456.1 | - | 7 | 764 | |||
AK316004.1 | - | 7 | 882 | |||
BC038582.1 | - | 7 | 773 | |||
BC050554.1 | - | 7 | 811 | 6202841 | ||
U49928.1 | - | 7 | 769 |